Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Rapid Ornithine
SDP

Catalog No. 23001800 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Fill Volume:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
23-001-800 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-800 Supplier Hardy Diagnostics™ Supplier No. K279
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid Ornithine can be used to detect ornithine decarboxylase activity in as little as two to four hours. This test will assist in the identification of Enterobacteriaceae and Staphylococcus lugdunensis.

  • The test can be used to determine ornithine decarboxylase activity in the family Enterobacteriaceae and can be used to differentiate Staphylococcus lugdunensis from other species of Staphylococcus
  • Especially useful to differentiate Proteus mirabilis from other Proteus spp., which are ornithine decarboxylase negative
  • Decarboxylase basal media consists of peptones and yeast extract which supply nitrogenous and other nutrients necessary for bacterial growth.
  • Bromcresol purple is a pH indicator
  • The amino acid ornithine is added to detect the production of the enzyme ornithine decarboxylase

Specifications

Appearance Clear
Broth Type Decarboxylase Basal Media
Certifications/Compliance GSA, ISO 13485, ISO 17025 , ANAB Calibration ISO 17025, BSI-ISO 13485
Color Pale Yellow to Light Grayish-purple
Content And Storage 2°C to 8°C
Additive Bromcresol Purple
Detects Ornithine decarboxylase Activity, Enterobacteriaceae and Staphylococcus lugdunensis
Format Tube
Manufacturing Origin United States
Media Type Microbiological Culture Media
Product Type Decarboxylase medium base
Test Type Microbial Identification
Fill Volume 1 mL
Formulation L-Ornithine 10.0gm, Meat Peptone 5.0gm, Yeast Extract 3.0gm, Bromcresol Purple 1.0gm
For Use With (Application) Identification of Enterobacteriaceae and Staphylococcus lugdunensis
Incubation Conditions 35°C, Aerobic
Incubation Time 2 to 4 hr.
Material Polycarbonate
No. per Pack 20
Packaging Type Box
Quantity 20 x 1 mL
Transfer Volume 0.5 mL
Dimensions (Dia. x H) 13 x 100 mm
Height (Metric) 100 mm
Diameter (Metric) 13 mm
Species Staphylococcus species
Protein Source Meat Peptone
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.