Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Standard Diagnostics Rapid Plasma Reagin (RPR) Test Kit
SDP

Catalog No. 23700101
Encompass
Change view
Click to view available options
Content And Storage:
2°C to 8°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
23-700-101 2°C to 8°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-700-101 Supplier Gold Standard Diagnostics Supplier No. GSD011600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Antigen Suspension
  • Reactive Control
  • Non-reactive Control

Determines the presence of reagin antibodies in human serum. It may be used to aid in the diagnosis of syphilis when used in conjunction with supplemental treponemal laboratory tests and other clinical information. Not intended for the screening of blood or tissue donors.

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Syphilis Testing
Includes Antigen Suspension
Reactive Control
Non-reactive Control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.