Learn More
New England Biolabs, Inc. Rapid™ PNGase F – 50 reactions
SDP
Catalog No. NC0855354
Encompass
Rapid PNGase F enables complete and rapid deglycosylation of antibodies and immunoglobulin fusion proteins, as well as other glycoproteins, in minutes. All N-glycans are released rapidly and without bias, and are ready to be prepared for downstream chromatography or mass spectrometry analysis. Rapid PNGase F creates an optimized workflow, reducing processing time without compromising sensitivity or reproducibility.
  • Complete deglycosylation of antibodies and immunoglobulin fusion proteins in 10 minutes
  • Recombinant enzyme; guaranteed endoglycosidase F1, F2 or F3 free
  • All reaction components are compatible with HPLC and mass spectrometry analysis
  • >= 95% purity, as determined by SDS-PAGE and intact ESI-MS
  • Optimal activity and stability ensured for up to 12 months
Catalog No. NC0855354 Supplier New England Biolabs, Inc. Supplier No. P0710S
Rapid PNGase F enables complete and rapid deglycosylation of antibodies and immunoglobulin fusion proteins, as well as other glycoproteins, in minutes. All N-glycans are released rapidly and without bias, and are ready to be prepared for downstream chromatography or mass spectrometry analysis. Rapid PNGase F creates an optimized workflow, reducing processing time without compromising sensitivity or reproducibility.
  • Complete deglycosylation of antibodies and immunoglobulin fusion proteins in 10 minutes
  • Recombinant enzyme; guaranteed endoglycosidase F1, F2 or F3 free
  • All reaction components are compatible with HPLC and mass spectrometry analysis
  • >= 95% purity, as determined by SDS-PAGE and intact ESI-MS
  • Optimal activity and stability ensured for up to 12 months