Learn More
New England Biolabs, Inc. Rapid™ PNGase F (non-reducing format) – 50 reactions
SDP
Catalog No. NC1069509
Encompass
Developed for proteomic applications, Rapid PNGase F (non-reducing format) enables complete and rapid deglycosylation, while preserving disulfide bonds. This facilitates high throughput proteomics applications and methods for antibody characterization by mass spectrometry, such as intact mass analysis. Rapid PNGase F (non-reducing format) combines the advantages of Rapid PNGase F (fast processing time), with non-reducing conditions, preserving quaternary structure.
  • Complete deglycosylation of antibodies and immunoglobulin fusion proteins in 15 minutes
  • No reducing agents in the formulation, samples remain intact following deglycosylation
  • Recombinant enzyme; guaranteed endoglycosidase F1, F2 or F3 free
  • All reaction components are compatible with HPLC and mass spectrometry analysis
  • Optimal activity and stability for up to 12 months
Catalog No. NC1069509 Supplier New England Biolabs, Inc. Supplier No. P0711S
Developed for proteomic applications, Rapid PNGase F (non-reducing format) enables complete and rapid deglycosylation, while preserving disulfide bonds. This facilitates high throughput proteomics applications and methods for antibody characterization by mass spectrometry, such as intact mass analysis. Rapid PNGase F (non-reducing format) combines the advantages of Rapid PNGase F (fast processing time), with non-reducing conditions, preserving quaternary structure.
  • Complete deglycosylation of antibodies and immunoglobulin fusion proteins in 15 minutes
  • No reducing agents in the formulation, samples remain intact following deglycosylation
  • Recombinant enzyme; guaranteed endoglycosidase F1, F2 or F3 free
  • All reaction components are compatible with HPLC and mass spectrometry analysis
  • Optimal activity and stability for up to 12 months