Learn More
Electron Microscopy Sciences Rapid Print Washer
SDP
Catalog No. 5028259
Encompass_Preferred
Water from 21 powerful jets runs constantly to separate and wash both sides of prints quickly and completely. Agitating water engages and dilutes the hypo, carrying it through the washer and out the opposite side quickly. The Rapid Print Washer accommodates 10 to 12 full-sized prints in one loading; all size films up to 11x14" (28x35.5cm). Internal measures: 13"x15 1/2".
Catalog No. 50-282-59 Supplier Electron Microscopy Sciences Supplier No. 74387
Water from 21 powerful jets runs constantly to separate and wash both sides of prints quickly and completely. Agitating water engages and dilutes the hypo, carrying it through the washer and out the opposite side quickly. The Rapid Print Washer accommodates 10 to 12 full-sized prints in one loading; all size films up to 11x14" (28x35.5cm). Internal measures: 13"x15 1/2".