Learn More
Siemens Healthineers Rapid QC Plus Blood Gas Quality Control
Catalog No. 23041214 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
1
2
3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-041214 3
09-529-243 1
09-529-275 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-041214 Supplier Siemens Healthineers Supplier No. 10325104
Add to Cart
Add to Cart
  • 30 ampules per level
  • 2.5mL of organic salt buffered bicarbonate solution per ampule
  • Yields expected values for pH/blood gas, Na+, K+, Cl- and Ca+ + (ionized and total calcium)
TRUSTED_SUSTAINABILITY
Specifications
Type Blood Gas Quality Control
Intended Use Used for yielding expected values for pH/Blood gas, Na+, K+, Cl-, Ca++
Level 3
Quantity 30 x 2.5 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov