Learn More
Analytical Sales & Services Rapid Replacement Pod for VXH593-983 - Rheodyne SS 6 Position, 7 Port Ultra High Pressure Switching Valve
Catalog No. 5000805644
Encompass
Rapid Replacement Pod for VXH593-983 - Rheodyne Stainless Steel 6 Position, 7 Port Ultra High Pressure Switching Valve. 15,000psi (1034 bar )\n\nQty/Pkg: EACH
Catalog No. 50-008-05644 Supplier Analytical Sales & Services Supplier No. PD5939
Rapid Replacement Pod for VXH593-983 - Rheodyne Stainless Steel 6 Position, 7 Port Ultra High Pressure Switching Valve. 15,000psi (1034 bar )\n\nQty/Pkg: EACH