Learn More
BTNX Inc BTNX CLIA-Moderate Pregnancy Test Cassettes (Urine/Serum) Combo 25 Cassettes/Box hCG 20 mIU
Catalog No. NC1114909
Encompass_Preferred
The Rapid Response™ Pregnancy Combo Test Cassette is a rapid visual immunoassay for the qualitative, presumptive detection of human chorionic gonadotropin in human urine or serum specimens. This kit is intended for use as an aid in the early detection of pregnancy. Sensitivity: 20 mIU/mL. Quantity: 25/kit. Time to result: 3 minutes (urine), 5 minutes (serum). CLIA-Moderate.
This item is not returnable. View return policy
Catalog No. NC1114909 Supplier BTNX Inc Supplier No. HCG3C25
The Rapid Response™ Pregnancy Combo Test Cassette is a rapid visual immunoassay for the qualitative, presumptive detection of human chorionic gonadotropin in human urine or serum specimens. This kit is intended for use as an aid in the early detection of pregnancy. Sensitivity: 20 mIU/mL. Quantity: 25/kit. Time to result: 3 minutes (urine), 5 minutes (serum). CLIA-Moderate.