Learn More
BTNX Inc Rapid Response™ hCG Pregnancy Control Solutions 20 mIU
Catalog No. NC1114911
Encompass_Preferred
The Rapid Response™ 20 mIU hCG Pregnancy Control Solutions is to be used with the corresponding Rapid Response™ 20 mIU Pregnancy Test Kits as a qualitative control sample to verify the procedure and test performance for the Rapid Response™ 20 mIU Pregnancy Test Kits. The kit includes a positive control solution, a negative control solution, and instructions.
This item is not returnable. View return policy
Catalog No. NC1114911 Supplier BTNX Inc Supplier No. HCG1C20NP
The Rapid Response™ 20 mIU hCG Pregnancy Control Solutions is to be used with the corresponding Rapid Response™ 20 mIU Pregnancy Test Kits as a qualitative control sample to verify the procedure and test performance for the Rapid Response™ 20 mIU Pregnancy Test Kits. The kit includes a positive control solution, a negative control solution, and instructions.