Learn More
BTNX Inc Rapid Response™ Pregnancy Test Cassette - 50 Tests
Catalog No. NC1114908
Encompass_Preferred
The Rapid Response™ Pregnancy Test Cassette test for human chorionic gonadotropin (hCG) in human urine specimens, to aid in the early detection of pregnancy. Kit contains 50 Tests per kit. The Rapid Response™ Pregnancy Test Cassette is a rapid visual read for the qualitative, presumptive detection of hCG in urine specimens at 20 mIU/mL or higher. Although first morning urine specimens are preferable, urine specimens from any time of day can be used. Each test cassette is individually sealed in a pouch with a disposable pipette. “C” and “T” are printed directly on the cassette for easy-to-read test result lines. In just 3 minutes, test results can be observed through the visual interpretation of colored lines.
This item is not returnable. View return policy
Catalog No. NC1114908 Supplier BTNX Inc Supplier No. HCG1C50
The Rapid Response™ Pregnancy Test Cassette test for human chorionic gonadotropin (hCG) in human urine specimens, to aid in the early detection of pregnancy. Kit contains 50 Tests per kit. The Rapid Response™ Pregnancy Test Cassette is a rapid visual read for the qualitative, presumptive detection of hCG in urine specimens at 20 mIU/mL or higher. Although first morning urine specimens are preferable, urine specimens from any time of day can be used. Each test cassette is individually sealed in a pouch with a disposable pipette. “C” and “T” are printed directly on the cassette for easy-to-read test result lines. In just 3 minutes, test results can be observed through the visual interpretation of colored lines.