Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics Rapid SARS-CoV-2 Control Panel
SDP

Catalog No. 23264780 Shop All Microbiologics Products
Change view
Click to view available options
For Use With (Equipment):
Covid testing
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-264-780 Covid testing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-264-780 Supplier Microbiologics Supplier No. 8268
Add to Cart
Add to Cart

Rapid SARS-CoV-2 Control Panel contains six positive, inactivated control swabs of SARS-CoV-2 Virus.

Helix Elite QC Panels and Molecular Standards are FDA Class I exempt unassayed, in vitro diagnostic external controls, and are intended for professional laboratory use only. Results with diagnostic instruments may vary.

Specifications

Detectable Analytes SARS-CoV-2
Format Swab
For Use With (Equipment) Covid testing
Sample Type Swab
Type Control Panel
Disposable Single-use
Includes Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.