Learn More
Nacalai USA Inc RAPID Slit Seal (100 sheets/box) : Instant Self-closing Seal after Dispensing
Catalog No. nc1831702
Encompass
RAPID Slit Seal is a unique self-closing 96-well plate seal to minimize solvent evaporation and prevent needle clogging compared with conventional silicon mats. The adhesive-free, pre-cut slits made of silicone and PET allows easy insertion and withdrawal of pipette tips and sampling needles without catching and dragging. Acrylic-based adhesive is not above wells to prevent sample leaching. Seal can be easily applied and removed. Suggested applications include automated pipetting and liquid handling, SPE, HPLC, LC/MS, cell culturing and ADME.
Catalog No. NC1831702 Supplier Nacalai USA Inc Supplier No. R80.120.00
RAPID Slit Seal is a unique self-closing 96-well plate seal to minimize solvent evaporation and prevent needle clogging compared with conventional silicon mats. The adhesive-free, pre-cut slits made of silicone and PET allows easy insertion and withdrawal of pipette tips and sampling needles without catching and dragging. Acrylic-based adhesive is not above wells to prevent sample leaching. Seal can be easily applied and removed. Suggested applications include automated pipetting and liquid handling, SPE, HPLC, LC/MS, cell culturing and ADME.