Learn More
Description
Identify 40 different staphylococci and related genera in just four hours using the Thermo Scientific™ RapID™ STAPH PLUS System, an identification system based on enzyme technology.
- One-step inoculation—decreased prep time for increased productivity
- Manual microbial identification made easy—no oil, no pipetting, no 24-hr incubation
- Four-hour incubation*—quicker time-to-result for a faster response
- Common procedure—lessened materials usage saves time and money
- Visible color reactions—reduced subjectivity decreases repeat testing
- Better reporting—more coverage, advanced updates
A clear plastic tray contains reagent impregnated wells, allowing simultaneous inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Inoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.
After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a database or through the use of a computer-generated Code Compendium (ERIC).
Requires:
Requires RapID Nitrate A, (R8309003); RapID Nitrate B, (R8309004); RapID Inoculation Fluid, (R8325106).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | RapID STAPH PLUS System |
| CE Marker | Yes |
| Detectable Analytes | Staphylococcus species |
| Type | Staphylococcus aureus Test Kit |
| For Use With (Application) | Enzyme Immunoassay Detection |
| Quantity | 20 Panels/Kit |
| Species | Staphylococcus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
