Learn More
Nacalai USA Inc RAPID Tube Plate 600 (20 plates) : 96-well Plate Specialized for Metabolic Stability Assay in Drug Discovery
Catalog No. NC1831706
Encompass_Preferred
The RAPID Tube Plate 600 is ideal for in vitro metabolic stability studies. The enzyme assay plate is made of polypropylene and does not contain any enzyme inhibitors such as EDTA and other contaminants. In addition, superior thermal distribution between and among each well provides less variation in results after incubation. The 600 µL capacity per well allows flexibility in study design, and suggested applications are Kinetic Assays, High Throughput Screening, Cold Storage, and Process Automation.
Catalog No. NC1831706 Supplier Nacalai USA Inc Supplier No. R80.600.00
The RAPID Tube Plate 600 is ideal for in vitro metabolic stability studies. The enzyme assay plate is made of polypropylene and does not contain any enzyme inhibitors such as EDTA and other contaminants. In addition, superior thermal distribution between and among each well provides less variation in results after incubation. The 600 µL capacity per well allows flexibility in study design, and suggested applications are Kinetic Assays, High Throughput Screening, Cold Storage, and Process Automation.