Learn More
Protifi LLC S-Trap™ midi columns (≥ 300 μg) 40 pack
SDP
Catalog No. NC1679156
Encompass
The patent-pending S-Trap™ brings universal, simple and inexpensive sample preparation to bottom-up proteomics. One easy-to-use spin column combines sample concentration, clean up and digestion. S-Trap™ sample processing does not require any specialized equipment nor training. From µg to mg scales, your samples are ready in about an hour. S-Traps™ eliminate one of the greatest sources of variability – inconsistent lysis and protein solubilization – by using strong 5% SDS for all samples. This almost universal protein solvent is removed in the protein trap of the S-Trap™, which retains all undigested proteins but does not retain small molecules including the SDS used to extract and handle proteins.
Catalog No. NC1679156 Supplier Protifi LLC Supplier No. C02MIDI40
The patent-pending S-Trap™ brings universal, simple and inexpensive sample preparation to bottom-up proteomics. One easy-to-use spin column combines sample concentration, clean up and digestion. S-Trap™ sample processing does not require any specialized equipment nor training. From µg to mg scales, your samples are ready in about an hour. S-Traps™ eliminate one of the greatest sources of variability – inconsistent lysis and protein solubilization – by using strong 5% SDS for all samples. This almost universal protein solvent is removed in the protein trap of the S-Trap™, which retains all undigested proteins but does not retain small molecules including the SDS used to extract and handle proteins.