Learn More
StatLab RapidCal Decalcifier
SDP
Catalog No. 21033252
Change view
Click to view available options
Solution Type:
Rapid Cal
Catalog No. Solution Type
21-033-252 Rapid Cal
1 options
Catalog No. 21-033-252 Supplier StatLab Supplier No. 6040
Add to Cart
Add to Cart
Formulated to decalcify bone rapidly
  • Formulated to decalcify bone rapidly, yet perform gently enough to be used as a routine decalcifying solution for bone marrow biopsies
  • Designed to preserve maximum immogenicity while accomplishing rapid decalcification
  • 4 gallons per case
Specifications
Product Type Decalcifying Solution
Solution Type Rapid Cal
Form Solution