Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab RapidFix
SDP

Catalog No. 21033251
Change view
Click to view available options
Capacity:
15 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
21-033-251 15 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-251 Supplier StatLab Supplier No. 0175
Add to Cart
Add to Cart

A buffered formalin fixative for use at the grossing station

  • Fixes approximately 50% faster than 10% NBF
  • Safe enough for use on all tissue specimens
  • Compatible with all other primary fixatives with no interference with any special stain or IHC procedures

Specifications

Capacity 15 gal.
For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Formalin
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.