Learn More
Siemens Healthineers RAPIDLab™ General Chemistry High Glucose/Lactate Reagent
Catalog No. 23044832 Shop All Siemens Healthineers Products
Change view
Click to view available options
Type:
General Chemistry Reagent
Catalog No. Type
23-044-832 General Chemistry Reagent
1 options
This item is not returnable. View return policy
Catalog No. 23-044-832 Supplier Siemens Healthineers Supplier No. 10311387
Add to Cart
Add to Cart
This item is not returnable. View return policy
For in vitro diagnostic use for verifying glucose and lactate sensor performance on the RAPIDLab™ 1260 & 1265 systems.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) RAPIDLab™ 1260 & 1265 systems
Type General Chemistry Reagent
For Use With (Application) Verify glucose and lactate sensor performance