Learn More
Siemens Healthineers RAPIDLyte Multicap™-S Plastic Blood Collection Capillary Tubes
Catalog No. 23044855 Shop All Siemens Healthineers Products
Change view
Click to view available options
Volume (Metric):
100 μL
140 μL
175 μL
Quantity:
50 Tubes
500 Tubes
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
23-044-855 175 μL 50 Tubes
23-044-839 100 μL 50 Tubes
23-044-851 100 μL 500 Tubes
23-044-843 140 μL 50 Tubes
23-044-838 140 μL 500 Tubes
23-044-842 175 μL 500 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 23-044-855 Supplier Siemens Healthineers Supplier No. 10324363
Add to Cart
Add to Cart
This item is not returnable. View return policy
Multicap™-S capillaries are designed to minimize pre-analytical error, keep users safe and maximize workflow efficiencies.
Multicap™-S capillary tubes are available in a variety of sizes and configurations to meet your clinical needs across a range of critical care settings.
  • Choose from a variety of capillaries in multiple configurations to meet your arterial blood sampling needs
  • Maintain the highest level of sample integrity with no dilution errors, minimized electrolyte binding and fast, easy air bubble removal
  • Maximize your instrument uptime with our advanced, dry-sprayed, balanced and reduced heparin formulations to minimize the potential for clots
TRUSTED_SUSTAINABILITY
Specifications
Product Type Blood Collection Capillary Tubes
Material Plastic
Volume (Metric) 175 μL
For Use With (Application) pH/Blood Gas/Na+/K+/Ca++/Cl-/Glu/Lac/tHb Testing
Quantity 50 Tubes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov