Learn More
PROTEIOS TECHNOLOGY INC RapidPro™ FPLC 1 mL Kit
Catalog No. NC3591635
Encompass
The RapidPro™ FPLC 1 mL Kit uses Car9-tag affinity for high-performance purification, achieving >90% purity in 30 min. Compatible with ÄKTA™ systems and syringe pumps for flexible protein expression workflows.
Catalog No. NC3591635 Supplier PROTEIOS TECHNOLOGY INC Supplier No. SQ1589560
The RapidPro™ FPLC 1 mL Kit uses Car9-tag affinity for high-performance purification, achieving >90% purity in 30 min. Compatible with ÄKTA™ systems and syringe pumps for flexible protein expression workflows.