Learn More
PROTEIOS TECHNOLOGY INC RapidPro™ FPLC 5 mL Kit
Catalog No. NC3591637
Encompass
The RapidPro™ FPLC 5 mL Kit uses Car9-tag affinity for high-yield purification, achieving >90% purity in 40 min. Its 5 mL resin bed supports larger lysate volumes with excellent binding efficiency and reproducibility.
Catalog No. NC3591637 Supplier PROTEIOS TECHNOLOGY INC Supplier No. SQ7611433
The RapidPro™ FPLC 5 mL Kit uses Car9-tag affinity for high-yield purification, achieving >90% purity in 40 min. Its 5 mL resin bed supports larger lysate volumes with excellent binding efficiency and reproducibility.