Learn More
PROTEIOS TECHNOLOGY INC RapidPro™ Minispin Kit
Catalog No. NC3591623
Encompass
The RapidPro™ MiniSpin Kit uses Car9-tag affinity for fast, high-purity protein purification in 25 min, integrating with microcentrifuges (6000 x g) and single-use spin columns for small-volume applications.
Catalog No. NC3591623 Supplier PROTEIOS TECHNOLOGY INC Supplier No. SQ2903758
The RapidPro™ MiniSpin Kit uses Car9-tag affinity for fast, high-purity protein purification in 25 min, integrating with microcentrifuges (6000 x g) and single-use spin columns for small-volume applications.