Learn More
PROTEIOS TECHNOLOGY INC RapidPro™ Spin Kit
Catalog No. NC3591622
Encompass
The RapidPro™ Spin Kit uses Car9-tag affinity for fast, high-purity protein purification in 20 min, processing up to 4 mL of lysate with centrifuge-compatible spin columns for mid-scale purification.
Catalog No. NC3591622 Supplier PROTEIOS TECHNOLOGY INC Supplier No. SQ3428749
The RapidPro™ Spin Kit uses Car9-tag affinity for fast, high-purity protein purification in 20 min, processing up to 4 mL of lysate with centrifuge-compatible spin columns for mid-scale purification.