Learn More
Electron Microscopy Sciences Rapid Pro Tissue Processing Solution 1 250 ML
SDP
Catalog No. 501929635
Encompass_Preferred
The EMS Rapid Pro Processing System is a proven and reliable method for obtaining rapid results in tissue processing. It can cut total processing time to as little as 46 minutes. In addition to fast processing, the system has many advantages over traditional processing.
Increased throughput.
Suitable for both fresh and frozen sections.
Catalyzed versions of standard processing chemicals eliminate the need for new safety features, storage, or learning curves.
Effective for manual or automatic tissue processing.
Compatible with existing tissue processing equipment.
Catalog No. 50-192-9635 Supplier Electron Microscopy Sciences Supplier No. 2675001
The EMS Rapid Pro Processing System is a proven and reliable method for obtaining rapid results in tissue processing. It can cut total processing time to as little as 46 minutes. In addition to fast processing, the system has many advantages over traditional processing.
Increased throughput.
Suitable for both fresh and frozen sections.
Catalyzed versions of standard processing chemicals eliminate the need for new safety features, storage, or learning curves.
Effective for manual or automatic tissue processing.
Compatible with existing tissue processing equipment.