Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RapidPure™ DNA Tissue Kit

Catalog No. MP112712050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
50 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP112712050 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112712050 Supplier MP Biomedicals™ Supplier No. 112712050

missing translation for 'validMainframeMsg'

Designed to isolate and purify total DNA from fresh, frozen or lyophilized plant material (such as leaves, roots, fruits or seeds) and also from a wide variety of food samples from plant origin (fresh, frozen or dried material)

Applications: up to 50 µg DNA from 100 mg plant tissue
Product Description: The RapidPURE DNA Plant Kit is designed to isolate and purify total DNA from fresh, frozen or lyophilized plant material (such as leaves, roots, fruits or seeds) and also from a wide variety of food samples from plant origin (fresh, frozen or dried material). The RapidPURE DNA Plant Kit is the ideal tool for a rapid and efficient isolation of high quality genomic DNA from up to 100 mg plant material
Recommended Use: isolate and purify total DNA from fresh, frozen or lyophilized plant material and also from a wide variety of food samples from plant origin
TRUSTED_SUSTAINABILITY

Specifications

Concentration up to 100 mg plant tissue
Content And Storage Store at Room temperature (15-30 °C).
Sample Type Plant Material, Food Samples from Plant Origin (Fresh, Frozen or Dried)
For Use With (Application) Nucleic Acid Isolation & Purification
Includes Plant Lysis Buffer, Binding Buffer, Proteinase K, Wash Buffer 1, Wash Buffer 2, Elution Buffer, Prefilter, Spin Filter, 2.0 ml Collection Tubes, 1.5 ml Collection Tubes, Manual
No. of Reactions 50 Preps
Product Type Kits & Assays
Form Chemical Lysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.