Learn More
Boca Scientific Inc RapiDri Integrated DNA Buccal Swab Kit, Swab with Stabilization Pouch and Bar Code, Pack of 200
Catalog No. NC1894473
Encompass
The RapiDri Swab kit contains the original high yield Isohelix DNA buccal swabs in combination with the new moisture wicking RapiDri Pouch and barcoded sealing label. Once placed in the RapiDri pouch, the swab is quickly stabilized to maximize the DNA yield without loss of integrity. Stabilization lasts for 3-6 months at ambient temperatures. The RapiDri pouch also protects the DNA from cross contamination and acts as a transport pack, meeting most international shipping requirements. This kit provides an economic option for shorter term buccal DNA stabilization with the swab and stabilization method conveniently boxed together. 200 swabs with the RapiDri Pouch and barcoded sealing label are included.
Catalog No. NC1894473 Supplier Boca Scientific Inc Supplier No. RD01
The RapiDri Swab kit contains the original high yield Isohelix DNA buccal swabs in combination with the new moisture wicking RapiDri Pouch and barcoded sealing label. Once placed in the RapiDri pouch, the swab is quickly stabilized to maximize the DNA yield without loss of integrity. Stabilization lasts for 3-6 months at ambient temperatures. The RapiDri pouch also protects the DNA from cross contamination and acts as a transport pack, meeting most international shipping requirements. This kit provides an economic option for shorter term buccal DNA stabilization with the swab and stabilization method conveniently boxed together. 200 swabs with the RapiDri Pouch and barcoded sealing label are included.