Description
- Plastic handheld design supports convenient, flexible sealing away from the base unit
- Helps extend the useful life of existing RapidSeal II equipment
- Supports fast RF sealing of PVC tubing in blood processing and laboratory workflows
- Ideal for replacing worn or damaged handheld sealing components
- Designed for use with compatible Genesis RapidSeal II systems
Specifications
Specifications
| Material | Plastic |
| Product Type | Handheld Sealer |
| For Use With (Application) | Tube Sealing |
| For Use With (Equipment) | SE340, SE540, SE640 |
| Includes | Replacement Handheld Sealer |
| Quantity | 1 Ea. |