Learn More
Genesis BPS™ RapidSeal II SE540 Tube Sealer
Catalog No. 22050486 Shop All Globe Scientific Products
Change view
Click to view available options
Size:
6 mm (0.25 in.)
Catalog No. Size
22-050-486 6 mm (0.25 in.)
1 options
Catalog No. 22-050-486 Supplier Genesis BPS™ Supplier No. BPS43CCS2017
Add to Cart
Add to Cart
Pairs flexible multi-head sealing with handheld capability for versatile RF sealing of PVC tubing
  • Supports multiple sealing workflows from a single system configuration
  • Handheld sealer improves reach and flexibility around bags and tubing sets
  • Designed for fast, repeatable sealing of PVC tubing
  • Built for blood processing, laboratory, and plasma handling applications
  • Ideal where both benchtop consistency and handheld flexibility are needed
Specifications
Product Type Multi-head Flexible Tube Sealer
Includes Fixed and Handheld Sealer
Size 6 mm (0.25 in.)