Learn More
Genesis BPS™ RapidSeal II Tube Sealer SE330
Catalog No. 22050477 Shop All Globe Scientific Products
Change view
Click to view available options
Size:
6 mm (0.25 in.)
Catalog No. Size
22-050-477 6 mm (0.25 in.)
1 options
Catalog No. 22-050-477 Supplier Genesis BPS™ Supplier No. BPS4282000481
Add to Cart
Add to Cart
Benchtop RF tube sealer with a fixed sealing head
  • Designed for fast sealing of PVC tubing in blood processing and laboratory applications
  • Consistent sealing performance supports high-throughput and routine workflows
  • Compact format fits efficiently on laboratory benches and processing areas
  • Simple operation supports repeatable seals with operator confidence
  • Ideal for blood banks, plasma centers, clinical laboratories, and processing facilities
Specifications
Product Type Tube Sealer
Includes Fixed Sealing Head
Size 6 mm (0.25 in.)