Learn More
Genesis BPS™ RapidSeal II Tube Sealer SE530
Catalog No. 22050462 Shop All Globe Scientific Products
Change view
Click to view available options
Size:
6 mm (0.25 in.)
Catalog No. Size
22-050-462 6 mm (0.25 in.)
1 options
Catalog No. 22-050-462 Supplier Genesis BPS™ Supplier No. BPS3BT10
Add to Cart
Add to Cart
Flexible multi-head RF tube sealer with a fixed sealing head, engineered for consistent PVC tube sealing
  • Multi-head design supports efficient processing and repeatable seal placement
  • Compact benchtop format fits blood bank and laboratory environments
  • Supports routine and higher-volume sealing applications
  • Ideal for blood processing, plasma handling, and clinical laboratory workflows
Specifications
Product Type Multi-head Flexible Tube Sealer
Includes Fixed Sealing Head
Size 6 mm (0.25 in.)