Learn More
Genesis BPS™ RapidSeal II Tube Sealer SE640
Catalog No. 22050466 Shop All Globe Scientific Products
Change view
Click to view available options
Size:
6 mm (0.25 in.)
Catalog No. Size
22-050-466 6 mm (0.25 in.)
1 options
Catalog No. 22-050-466 Supplier Genesis BPS™ Supplier No. BPS42040205
Add to Cart
Add to Cart
Flexible multi-head RF tube sealer designed for high-performance PVC tube sealing in blood processing environments
  • Supports efficient, repeatable seal placement in demanding workflows
  • Designed for fast sealing performance in blood bank and laboratory applications
  • Multi-head configuration helps improve throughput and processing consistency
  • Durable benchtop design supports routine use in high-volume environments
  • PVC tubing sealing for cell therapy, biopharmaceutical applications
  • Ideal for blood processing, plasma handling, and clinical laboratory operations
Specifications
Product Type Handheld Sealer
For Use With (Application) Blood Processing, Cell Therapy, Biopharmaceutical Applications
Includes Portable Handheld Sealer
Size 6 mm (0.25 in.)