Learn More
Genesis BPS™ RapidSeal II Tube Sealer
Catalog No. 22050478 Shop All Globe Scientific Products
Change view
Click to view available options
Size:
6 mm (0.25 in.)
Catalog No. Size
22-050-478 6 mm (0.25 in.)
1 options
Catalog No. 22-050-478 Supplier Genesis BPS™ Supplier No. BPS428SE330
Add to Cart
Add to Cart
Combines a fixed sealing head with a handheld sealer for flexible RF sealing in blood processing and laboratory workflows
  • Supports flexible sealing of PVC tubing at the bench or directly at the product location
  • Designed for fast, consistent seals in blood processing and laboratory settings
  • Handheld option improves reach and workflow flexibility
  • Built for routine use in demanding processing environments
  • Ideal for blood banks, plasma centers, and clinical laboratory applications
Specifications
Product Type Dual Tube Sealer
Includes Fixed and Handheld Sealer
Size 6 mm (0.25 in.)