Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genesis BPS™ RapidSeal Pro™ SE9 Benchtop Tube Sealer

Catalog No. 22021866 Shop All Globe Scientific Products
Change view
Click to view available options
Size:
DxWxH: 14.3 x 2.6 x 6.6 in. (365 x 65 x 167 mm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-021-866 DxWxH: 14.3 x 2.6 x 6.6 in. (365 x 65 x 167 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-021-866 Supplier Genesis BPS™ Supplier No. BPS428SE9
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

It delivers dependable RF tube sealing for demanding blood, plasma, and processing workflows.

  • Built for high-throughput environments: The rugged benchtop platform supports busy facilites that require consistent sealing performance throughout daily operations
  • Optional multi-head configuration: The SE9 can be configured with optional multi-head capability to help increase throughput, improve workflow efficiency, and support scalable sealing operations
  • Precision seal performance: RF sealing technology supports repeatable, consistent seals, helping operators work with confidence across critical applications
  • Durable platform construction: It is designed for long-term use in demanding healthcare and processing environments, helping reduce downtime and support operational continuity
  • Optional metal hand sealing unit: Facilities can add the RapidSeal Pro™ Metal Hand Sealing Unit as an optional SE9 accessory, providing additional flexibility for handheld sealing applications
  • Supports standardized workflows: It helps simplify training, improve process consistency, and support repeatable operation across users and departments
  • Ideal for centralized sealing areas: The benchtop design is well suited for processing benches, laboratory workstations, blood component areas, and plasma processing environments
  • Designed to improve productivity: With dependable performance, optional scalability, and rugged construction, it helps facilities maintain efficient sealing workflows with confidence

Specifications

Material Metal
Product Type Handheld Sealer
For Use With (Application) PVC Tube Sealing
For Use With (Equipment) PVC Tubing
Includes Spare Fuses (2), Spare Splashguards (5), Link Bars (2), Thumb Screws (4), Signal Cable (1), Two-Unit Power Cable AC Power Cord
No. of Places Linkable multi-head configurations
Quantity 1 Ea.
Size DxWxH: 14.3 x 2.6 x 6.6 in. (365 x 65 x 167 mm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.