Description
- Designed for flexible handheld RF sealing of PVC tubing
- Supports cord blood, cellular therapy, and laboratory processing workflows
- Helps maintain uptime by replacing worn or damaged handheld sealing components
- Supports accurate, repeatable seals when paired with compatible SE470 equipment
- Useful accessory for facilities operating SE470 handheld sealing systems
Specifications
Specifications
| Material | Plastic |
| Product Type | Handheld Sealer |
| For Use With (Application) | Cord Blood Tube Sealing |
| For Use With (Equipment) | SE470, UCBJIG |
| Includes | Elongated Replacement Handheld Sealer |
| Quantity | 1 Ea. |