Learn More
Genesis BPS™ RapidSeal Tube Sealer SE470
Catalog No. 22050461 Shop All Globe Scientific Products
Change view
Click to view available options
Size:
6 mm (0.25 in.)
Catalog No. Size
22-050-461 6 mm (0.25 in.)
1 options
Catalog No. 22-050-461 Supplier Genesis BPS™ Supplier No. BPS3BT06
Add to Cart
Add to Cart
Handheld RF tube sealer designed for cord blood and cellular therapy applications requiring flexible, reliable PVC tube sealing
  • Handheld RF tube sealer designed for cord blood and cellular therapy workflows
  • Supports fast, reliable sealing of PVC tubing in controlled processing applications
  • Flexible handheld design allows sealing close to bags, tubing sets, or processing areas
  • Designed to support workflows where operator access and mobility matter
  • Compact format helps integrate sealing into space-limited laboratory environments
  • Ideal for cord blood banks, cell therapy labs, and clinical processing teams
Specifications
Product Type Handheld Sealer
Includes Elongated Handheld Sealer
Size 6 mm (0.25 in.)