Description
- Replacement blade module for the Genesis RapidWeld II STW Sterile Tube Welder
- 500-weld capacity supports larger workflows and scheduled maintenance programs
- Precision blade design helps maintain consistent tube cutting and weld preparation
- Fast module replacement supports uptime and operator efficiency
- Designed specifically for RapidWeld II STW waferless sterile welding systems
- Ideal for blood bank, cell therapy, and biopharma facilities with recurring welding volume
Specifications
Specifications
| Material | Stainless Steel |
| Product Type | Blade Module |
| For Use With (Application) | Sterile Tube Welding |
| For Use With (Equipment) | RapidWeld II STW Sterile Tube Welder |
| Includes | RapidWeld Blade Module, 500 Welds |
| Quantity | 500 Welds |
| Size | 500 Welds |