Description
- Precision stainless steel blade supports consistent weld preparation
- 250-weld capacity supports planned maintenance and workflow control
- Designed for fast, simple replacement to help minimize downtime
- Helps maintain reliable waferless sterile welding performance
- Essential accessory for facilities operating RapidWeld II STW systems
Specifications
Specifications
| Material | Stainless Steel |
| Product Type | Blade Module |
| For Use With (Application) | Sterile Tube Welding |
| For Use With (Equipment) | RapidWeld II STW Sterile Tube Welder |
| Includes | RapidWeld Blade Module, 250 Welds |
| Quantity | 250 Welds |