Description
- 1000-weld capacity supports high-volume facilities and planned maintenance cycles
- Precision blade supports clean tube preparation and repeatable weld performance
- Simple replacement process helps reduce downtime during active workflows
- Designed to maintain performance in waferless sterile tube welding applications
- Ideal for high-throughput blood banking, cell therapy, and biopharma operations
Specifications
Specifications
| Material | Stainless Steel |
| Product Type | Blade Module |
| For Use With (Application) | Sterile Tube Welding |
| For Use With (Equipment) | RapidWeld II STW Sterile Tube Welder |
| Includes | RapidWeld Blade Module, 1000 Welds |
| Quantity | 1000 Welds |