Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genesis BPS™ RapidWeldII STW Sterile Tube Welder

Catalog No. 22050472 Shop All Globe Scientific Products
Change view
Click to view available options
Material:
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
22-050-472 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-050-472 Supplier Genesis BPS™ Supplier No. BPS425CM735AN6B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delivers fast, waferless sterile PVC tube welding for blood banking, cell therapy, and biopharma workflows

  • Waferless sterile tube welder designed for closed-system PVC tubing connections
  • Eliminates disposable copper wafers to reduce consumable costs and waste
  • Creates strong, repeatable welds in approximately 15 sec.
  • Supports wet-to-wet, dry-to-dry, and mixed PVC tubing combinations
  • Parallel tube loading and guided indicators help simplify operator setup
  • Color touchscreen, alarms, and multilingual interface support controlled use
  • Ideal for blood processing, cell therapy, biopharma, and regulated laboratory workflows

Specifications

Material Stainless Steel
Product Type Sterile Tube Welder
For Use With (Application) Sterile Welding of PVC Tubing
For Use With (Equipment) RapidWeld II STW Sterile Tube Welder
Includes Sterile Tube Welder, Power Supply
Size Outer Diameter 3.98 to 4.88 mm, Inner Diameter 2.91 to 3.23 mm, Wall Thickness 0.55 to 0.85 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.