Learn More
LGC GENOMICS LLC (Lucigen) RapiDxFire Thermostable Reverse Transcriptase, 50 ReactionsA truly thermostable reverse transcriptase for fast synthesis of short cDNA (< 1 Kb).
Catalog No. 502231954
Encompass
RapiDxFire Thermostable Reverse Transcriptase, 50 ReactionsA truly thermostable reverse transcriptase for fast synthesis of short cDNA (< 1 Kb).Key features:Extremely active at high temperatures (55 ¿C to 80 ¿C): Improves specificity of cDNA Synthesis from diverse RNA templatesSensitive: Detects 100 copies of RNA in two-step RT-qPCR assaysShort reaction time (5 minutes or less): Streamlined RT-qPCR workflow and faster time to resultsStable at room temperature (> 3 months): Simplifies setup on automation decks (no cold storage required) and enables use in environments with limited cold-storageLyo-compatible: Enzyme formulation is free of glycerol -and other components- that are known to interfere with downstream lyophilisationBatch to batch reproducibility: Manufactured in an ISO 13485-certified facility
This item is not returnable. View return policy
Catalog No. 50-223-1954 Supplier LGC GENOMICS LLC Supplier No. 302501
RapiDxFire Thermostable Reverse Transcriptase, 50 ReactionsA truly thermostable reverse transcriptase for fast synthesis of short cDNA (< 1 Kb).Key features:Extremely active at high temperatures (55 ¿C to 80 ¿C): Improves specificity of cDNA Synthesis from diverse RNA templatesSensitive: Detects 100 copies of RNA in two-step RT-qPCR assaysShort reaction time (5 minutes or less): Streamlined RT-qPCR workflow and faster time to resultsStable at room temperature (> 3 months): Simplifies setup on automation decks (no cold storage required) and enables use in environments with limited cold-storageLyo-compatible: Enzyme formulation is free of glycerol -and other components- that are known to interfere with downstream lyophilisationBatch to batch reproducibility: Manufactured in an ISO 13485-certified facility