Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Glycan Performance Test Standard, N-Glycan, RapiFluor-MS, 400 pmoles

Catalog No. NC1365269 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This RapiFluor-MS Glycan Performance Test Standard is a benchmarking standard and labeled, complex control for N-Glycan analysis. It contains N-glycan content from 30 μg of human IgG, providing approximately 400 pmoles of RapiFluor-MS labeled, released N-glycans.

  • Functions as a benchmarking standard and labeled control.
  • Contains N-glycan content from 30 μg of human IgG, approx. 400 pmoles of RapiFluor-MS labeled, released N-glycans.
  • Maximizes information from ms detection.
  • Streamlines deglycosylation and labeling.
  • Balances speed and sensitivity for confident analytical results.
  • High sensitivity mass detection for flr and ms.
  • Reduces chromatography analysis times.
  • Integrates with gmp-compliant uplc workflows.
  • Storage: –20°C upon arrival; –80°C (up to 3 months) or 4–10°C (up to one week) once solubilized.

Catalog No. NC1365269 Supplier Waters Corp Supplier No. 186007983
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This RapiFluor-MS Glycan Performance Test Standard is a benchmarking standard and labeled, complex control for N-Glycan analysis. It contains N-glycan content from 30 μg of human IgG, providing approximately 400 pmoles of RapiFluor-MS labeled, released N-glycans.

  • Functions as a benchmarking standard and labeled control.
  • Contains N-glycan content from 30 μg of human IgG, approx. 400 pmoles of RapiFluor-MS labeled, released N-glycans.
  • Maximizes information from ms detection.
  • Streamlines deglycosylation and labeling.
  • Balances speed and sensitivity for confident analytical results.
  • High sensitivity mass detection for flr and ms.
  • Reduces chromatography analysis times.
  • Integrates with gmp-compliant uplc workflows.
  • Storage: –20°C upon arrival; –80°C (up to 3 months) or 4–10°C (up to one week) once solubilized.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.