Learn More
Waters Corp Waters Corporation RapiFluor-MS Intact mAb Standard, Glycan, 400 pmol/vial
Catalog No. 502224213 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The RapiFluor-MS Intact mAb Standard is designed for method familiarization, troubleshooting, and benchmarking in glycan analysis and labeling workflows. This standard contains RapiFluor-MS labeled, released N-glycans obtained from 30 μg of Intact mAb Mass Check Standard and provides a concentration of 400 pmol/vial. It is specifically intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit.
  • Designed for method familiarization, troubleshooting, and benchmarking.
  • Contains RapiFluor-MS labeled, released N-glycans.
  • Obtained from 30 μg of Intact mAb Mass Check Standard.
  • Contains 400 pmol/vial.
  • Intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit.
  • Essential for understanding the workflow or verifying system performance.
  • Undergoes strict QC processes to guarantee product quality.
Catalog No. 50-222-4213 Supplier Waters Corp Supplier No. 186008843
The RapiFluor-MS Intact mAb Standard is designed for method familiarization, troubleshooting, and benchmarking in glycan analysis and labeling workflows. This standard contains RapiFluor-MS labeled, released N-glycans obtained from 30 μg of Intact mAb Mass Check Standard and provides a concentration of 400 pmol/vial. It is specifically intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit.
  • Designed for method familiarization, troubleshooting, and benchmarking.
  • Contains RapiFluor-MS labeled, released N-glycans.
  • Obtained from 30 μg of Intact mAb Mass Check Standard.
  • Contains 400 pmol/vial.
  • Intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit.
  • Essential for understanding the workflow or verifying system performance.
  • Undergoes strict QC processes to guarantee product quality.