Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION RapiGest SF Surfactant, 1 mg, 1/pk

Catalog No. 50780781 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

RapiGest SF is a patented anionic surfactant that enhances in-solution protein digestion speed and peptide recovery, particularly for mass spectrometry (MS) sample preparation. It avoids high background noise often seen with other surfactants and is provided as a 1 mg solid vial.

  • Drastically improves in-solution protein digestion speed and peptide recovery.
  • Valuable for mass spectrometry (MS) sample preparation by avoiding high background noise.
  • One, 1 mg vial provides material for 1 to 10 digestions.
  • Enhances digestion speed and increases peptide recovery when added to proteolytic digestion.
  • Formulation: Solid; Storage: Room temperature.
  • Shelf life: 36 months; Shelf life upon reconstitution: 1 week.
  • Shipping condition: Ambient; Application: Protein.

Catalog No. 50-780-781 Supplier Waters Corp Supplier No. 186001860
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RapiGest SF is a patented anionic surfactant that enhances in-solution protein digestion speed and peptide recovery, particularly for mass spectrometry (MS) sample preparation. It avoids high background noise often seen with other surfactants and is provided as a 1 mg solid vial.

  • Drastically improves in-solution protein digestion speed and peptide recovery.
  • Valuable for mass spectrometry (MS) sample preparation by avoiding high background noise.
  • One, 1 mg vial provides material for 1 to 10 digestions.
  • Enhances digestion speed and increases peptide recovery when added to proteolytic digestion.
  • Formulation: Solid; Storage: Room temperature.
  • Shelf life: 36 months; Shelf life upon reconstitution: 1 week.
  • Shipping condition: Ambient; Application: Protein.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.