Learn More
Waters Corp WATERS CORPORATION RapiGest SF, 10 mg, Protein Digestion Surfactant
Catalog No. 50780957 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
RapiGest™ SF is an innovative anionic surfactant that significantly improves in-solution protein digestion by increasing speed and peptide recovery. It is supplied as one 10 mg vial, suitable for larger-scale digestions, and enhances proteolytic digestion.
  • Enhances in-gel and in-solution enzymatic protein digestion, improving speed and peptide recovery without inhibiting enzyme activity.
  • Solubilizes proteins, including hydrophobic and enzyme-resistant types, making them more susceptible to cleavage.
  • Heat stable and acid labile, allowing for removal from solutions after hydrolysis without modifying peptides or suppressing endoprotease activity.
  • Accelerates peptide production by various proteases and reduces overall digestion time.
  • Degradation products do not interfere with LC-MS or MALDI MS analysis, avoiding high background noise in mass spectrometry.
Catalog No. 50-780-957 Supplier Waters Corp Supplier No. 186002123
RapiGest™ SF is an innovative anionic surfactant that significantly improves in-solution protein digestion by increasing speed and peptide recovery. It is supplied as one 10 mg vial, suitable for larger-scale digestions, and enhances proteolytic digestion.
  • Enhances in-gel and in-solution enzymatic protein digestion, improving speed and peptide recovery without inhibiting enzyme activity.
  • Solubilizes proteins, including hydrophobic and enzyme-resistant types, making them more susceptible to cleavage.
  • Heat stable and acid labile, allowing for removal from solutions after hydrolysis without modifying peptides or suppressing endoprotease activity.
  • Accelerates peptide production by various proteases and reduces overall digestion time.
  • Degradation products do not interfere with LC-MS or MALDI MS analysis, avoiding high background noise in mass spectrometry.