Learn More
Waters Corp Waters Corporation RapiGest SF, Anionic Surfactant, 3 mg, 1/pk
Catalog No. NC1272365 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
RapiGest SF is a patented anionic surfactant that significantly improves in-solution protein digestion speed and peptide recovery, especially valuable in protein sample preparation for mass spectrometry (MS) analysis by avoiding high background noise from other common surfactants.
  • Patented anionic surfactant
  • Improves in-solution protein digestion speed and peptide recovery
  • Avoids high background noise in mass spectrometry (MS) analysis
  • Enhances digestion speed and increases peptide recovery with various enzymes
  • Improves protein sequence coverage
  • Reduces sample preparation time and digestion time
  • Requires fewer enzymes for optimum digestion
  • Improves digestion of enzyme-resistant proteins
  • US Patent No: 8,580,533
Catalog No. NC1272365 Supplier Waters Corp Supplier No. 186008090
RapiGest SF is a patented anionic surfactant that significantly improves in-solution protein digestion speed and peptide recovery, especially valuable in protein sample preparation for mass spectrometry (MS) analysis by avoiding high background noise from other common surfactants.
  • Patented anionic surfactant
  • Improves in-solution protein digestion speed and peptide recovery
  • Avoids high background noise in mass spectrometry (MS) analysis
  • Enhances digestion speed and increases peptide recovery with various enzymes
  • Improves protein sequence coverage
  • Reduces sample preparation time and digestion time
  • Requires fewer enzymes for optimum digestion
  • Improves digestion of enzyme-resistant proteins
  • US Patent No: 8,580,533