Learn More
Waters Corp Waters RapiGest SF Reagent, 50 mg, 1 Pack
Catalog No. 50780956 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
RapiGest SF is an innovative anionic surfactant that significantly improves in-solution protein digestion speed and peptide recovery for mass spectrometry (MS) analysis by avoiding high background noise. This product contains 50 mg of the reagent, suitable for large-scale digestions.
  • Improves in-solution protein digestion speed and peptide recovery.
  • Avoids high background noise in mass spectrometry (MS) analysis.
  • Enhances enzymatic digestions of proteins.
  • Does not modify peptides or suppress endoprotease activity.
  • Easily removed after use by acidification for downstream analyses.
  • Supports high-temperature digestions due to heat stability.
Catalog No. 50-780-956 Supplier Waters Corp Supplier No. 186002122
RapiGest SF is an innovative anionic surfactant that significantly improves in-solution protein digestion speed and peptide recovery for mass spectrometry (MS) analysis by avoiding high background noise. This product contains 50 mg of the reagent, suitable for large-scale digestions.
  • Improves in-solution protein digestion speed and peptide recovery.
  • Avoids high background noise in mass spectrometry (MS) analysis.
  • Enhances enzymatic digestions of proteins.
  • Does not modify peptides or suppress endoprotease activity.
  • Easily removed after use by acidification for downstream analyses.
  • Supports high-temperature digestions due to heat stability.