Learn More
Waters Corp Proteinase K Digestion Module, Non-Specific, Enzymatic, Three-Reagent
Catalog No. 502886191 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The RapiZyme Proteinase K Digestion Module is a sample preparation kit for rapid, non-specific enzymatic degradation of proteins. It includes Proteinase K, guanidine HCl as a denaturant, and TCEP as a reductant, facilitating protein degradation in various assays like oligonucleotide extraction from biofluids and tissues, and degrading capsid proteins from viral vector samples.
  • Contains proteinase K, guanidine HCl, and TCEP.
  • Active between pH 4 and 12, with optimal activity at 50°C to 60°C.
  • Compatible with denaturants and surfactants.
  • Free of exonucleases, endonucleases, and ribonucleases.
  • Facilitates detergent-free digestion of proteins.
  • Aids in the disruption of oligonucleotide/protein binding.
  • Aids in viral vector digestion for genome integrity analysis.
  • Digestion can be achieved in as little as 40 min.
Catalog No. 50-288-6191 Supplier Waters Corp Supplier No. 186010601
The RapiZyme Proteinase K Digestion Module is a sample preparation kit for rapid, non-specific enzymatic degradation of proteins. It includes Proteinase K, guanidine HCl as a denaturant, and TCEP as a reductant, facilitating protein degradation in various assays like oligonucleotide extraction from biofluids and tissues, and degrading capsid proteins from viral vector samples.
  • Contains proteinase K, guanidine HCl, and TCEP.
  • Active between pH 4 and 12, with optimal activity at 50°C to 60°C.
  • Compatible with denaturants and surfactants.
  • Free of exonucleases, endonucleases, and ribonucleases.
  • Facilitates detergent-free digestion of proteins.
  • Aids in the disruption of oligonucleotide/protein binding.
  • Aids in viral vector digestion for genome integrity analysis.
  • Digestion can be achieved in as little as 40 min.