Learn More
NEOGEN Rappaport Vassiliadis R10 Broth, 10 kg
SDP
Catalog No. 502015482
Encompass_Preferred
Rappaport Vassiliadis R10 Broth is used for the selective enrichment of Salmonella spp. from foods in a laboratory setting. Rappaport Vassiliadis R10 Broth is not intended for use in the diagnosis of disease or other conditions in humans. Rappaport et al formulated an enrichment medium for Salmonella spp. that was modified by Vassiliadis et al. The Rappaport formulation, designated R 25/37°C, recommended incubation at 37°C. The Vassiliadis modification, designated R 10/43°C, had a reduced level of Malachite Green and recommended incubation at 43°C. Peterz later showed that incubation at 41.5 ± 0.5°C for 24 hours improved recovery of Salmonella spp. Rappaport-Vassiliadis R10 Broth is a selective enrichment medium that is used following pre-enrichment of the specimen.For more information visit https://www.neogen.com/solutions/Microbiology/rappaport-vassiliadis-r10-broth
Catalog No. 50-201-5482 Supplier NEOGEN Supplier No. 700003294
Rappaport Vassiliadis R10 Broth is used for the selective enrichment of Salmonella spp. from foods in a laboratory setting. Rappaport Vassiliadis R10 Broth is not intended for use in the diagnosis of disease or other conditions in humans. Rappaport et al formulated an enrichment medium for Salmonella spp. that was modified by Vassiliadis et al. The Rappaport formulation, designated R 25/37°C, recommended incubation at 37°C. The Vassiliadis modification, designated R 10/43°C, had a reduced level of Malachite Green and recommended incubation at 43°C. Peterz later showed that incubation at 41.5 ± 0.5°C for 24 hours improved recovery of Salmonella spp. Rappaport-Vassiliadis R10 Broth is a selective enrichment medium that is used following pre-enrichment of the specimen.For more information visit https://www.neogen.com/solutions/Microbiology/rappaport-vassiliadis-r10-broth