Learn More
NEOGEN Rappaport Vassiliadis Salmonella Enrichment Broth, 500 g
SDP
Catalog No. 502015450
Encompass_Preferred
Rappaport-Vassiliadis Salmonella Enrichment Broth is used for the enrichment and selective isolation of Salmonella spp. Conforms to Harmonized USP/EP/JP Requirements. Rappaport-Vassiliadis Salmonella Enrichment Broth is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized Pharmacopoeia for the enrichment and selective isolation of Salmonella spp.from non-sterile samples. Malachite green and magnesium chloride act as selective agents, combined with high osmotic pressure and low pH to effectively inhibit non-target competitive organisms. Soya peptone provides a source of nitrogen and potassium phosphate acts as a buffer. Some Salmonella (S. typhi and S. enteritidis) are known to be sensitive to malachite green and as such may fail to grow.For more information visit https://www.neogen.com/solutions/Microbiology/rappaport-vassiliadis-salmonella-enrichment-broth
Catalog No. 50-201-5450 Supplier NEOGEN Supplier No. 700003259
Rappaport-Vassiliadis Salmonella Enrichment Broth is used for the enrichment and selective isolation of Salmonella spp. Conforms to Harmonized USP/EP/JP Requirements. Rappaport-Vassiliadis Salmonella Enrichment Broth is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized Pharmacopoeia for the enrichment and selective isolation of Salmonella spp.from non-sterile samples. Malachite green and magnesium chloride act as selective agents, combined with high osmotic pressure and low pH to effectively inhibit non-target competitive organisms. Soya peptone provides a source of nitrogen and potassium phosphate acts as a buffer. Some Salmonella (S. typhi and S. enteritidis) are known to be sensitive to malachite green and as such may fail to grow.For more information visit https://www.neogen.com/solutions/Microbiology/rappaport-vassiliadis-salmonella-enrichment-broth