Learn More
Teknova Rappaport-Vassiliadis Broth (RVS). 10mL, 25-Pack, Sterile. 6 pack.
SDP
Catalog No. 502387272
Encompass
1.34% Magnesium Chloride, 0.72% Sodium Chloride, 0.45% Tryptone, 0.15% Monopotassium Phosphate, 36g/mL Malachite Green Oxalate
Catalog No. 50-238-7272 Supplier Teknova Supplier No. R02006PK
1.34% Magnesium Chloride, 0.72% Sodium Chloride, 0.45% Tryptone, 0.15% Monopotassium Phosphate, 36g/mL Malachite Green Oxalate